3-Bromo-2-hydroxy-5-(pentafluorothio)benzoic acid 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Bromo-2-hydroxy-5-(pentafluorosulphanyl)benzoic acid
PC56144-100MG
PC56144-250MG
APOSPC56144-100MGEA
145
EUR
APOSPC56144-100MG
APOSPC56144-250MG
3-Bromo-2-hydroxy-5-(pentafluorothio)benzoic acid 95+%
3-Bromo-2-hydroxy-5-(pentafluorothio)benzoic acid
Formel:
C₇H₄BrF₅O₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD31561290 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...