4-(2-Fluorophenyl)thiazol-2-ylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-(2-fluorophenyl)-1,3-thiazole

PC5615-1G
APOSPC5615-1GEA 58 EUR
APOSPC5615-1G
4-(2-Fluorophenyl)thiazol-2-ylamine
4-(2-Fluorophenyl)thiazol-2-ylamine
Formel: C₉H₇FN₂S
Lagertemperatur: Kühlschrank
MDL: MFCD02664014
CAS-Nummer: 145029-82-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR