2,3-Difluoro-4-iodobenzotrifluoride 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzene, 2,3-difluoro-1-iodo-4-(trifluoromethyl)-

PC56170-1G PC56170-250MG PC56170-5G
APOSPC56170-1GEA 281 EUR
APOSPC56170-1G APOSPC56170-250MG APOSPC56170-5G
2,3-Difluoro-4-iodobenzotrifluoride 95+%
2,3-Difluoro-4-iodobenzotrifluoride
Formel: C₇H₂F₅I
Lagertemperatur: Raumtemperatur
MDL: MFCD31568564
CAS-Nummer: 2227272-80-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR