2-Bromo-4-fluoro-3-iodobenzotrifluoride 97%

Lieferant: Apollo Scientific
Achtung

PC56173-1G PC56173-250MG
APOSPC56173-1GEA 1210 EUR
APOSPC56173-1G APOSPC56173-250MG
2-Bromo-4-fluoro-3-iodobenzotrifluoride 97%
2-Bromo-4-fluoro-3-iodobenzotrifluoride
Formel: C₇H₂BrF₄I
Lagertemperatur: Raumtemperatur
MDL: MFCD31568566
CAS-Nummer: 2227272-51-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR