(R)-1-(2, 4-Difluorophenyl)ethylamine

Lieferant: Apollo Scientific

Synonyme: (R)-1-(2,4-Difluorophenyl)ethanamine, (1R)-1-(2,4-difluorophenyl)ethanamine

PC56178-1G
APOSPC56178-1GEA 144 EUR
APOSPC56178-1G
(R)-1-(2, 4-Difluorophenyl)ethylamine
(R)-1-(2, 4-Difluorophenyl)ethylamine
Formel: C₈H₉F₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD06761833
CAS-Nummer: 791098-84-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR