(S)-1-(2, 4-Difluorophenyl)ethylamine 97%

Lieferant: Apollo Scientific

Synonyme: (1S)-1-(2,4-Difluorophenyl)ethanamine, (S)-1-(2,4-Difluorophenyl)ethanamine

PC56179-250MG
APOSPC56179-250MGEA 183 EUR
APOSPC56179-250MG
(S)-1-(2, 4-Difluorophenyl)ethylamine 97%
(S)-1-(2, 4-Difluorophenyl)ethylamine
Formel: C₈H₉F₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD06761834
CAS-Nummer: 845252-02-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR