2-Chloro-4-iodo-1-(trifluoromethyl)benzene

Lieferant: Apollo Scientific
Gefahr

PC56186-250MG
APOSPC56186-250MGEA 255 EUR
APOSPC56186-250MG
2-Chloro-4-iodo-1-(trifluoromethyl)benzene
2-Chloro-4-iodo-1-(trifluoromethyl)benzene
Formel: C₇H₃ClF₃I
Lagertemperatur: Kühlschrank
MDL: MFCD12827841
CAS-Nummer: 1206599-46-3
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR