3'-(Trifluormethyl)-2-phenylacetanilid
Lieferant: Apollo Scientific
|
Achtung
|
PC5621-5G
APOSPC5621-5GEA
205
EUR
APOSPC5621-5G
3'-(Trifluormethyl)-2-phenylacetanilid
3'-(Trifluormethyl)-2-phenylacetanilid
Formel:
C₁₅H₁₂F₃NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043457 CAS-Nummer: 1939-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...