5-Bromo-2-(trifluoromethoxy)anisole

Lieferant: Apollo Scientific
Achtung

PC56210-1G PC56210-250MG
APOSPC56210-1GEA 346 EUR
APOSPC56210-1G APOSPC56210-250MG
5-Bromo-2-(trifluoromethoxy)anisole
5-Bromo-2-(trifluoromethoxy)anisole
Formel: C₈H₆BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD21333068
CAS-Nummer: 672948-65-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR