1-Bromo-5-chloro-1,1,3,3,5,5-hexafluoropentane

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-5-chloro-1,1,3,3,5,5-hexafluoropentane

PC56223-1G
APOSPC56223-1GEA 87 EUR
APOSPC56223-1G
1-Bromo-5-chloro-1,1,3,3,5,5-hexafluoropentane
1-Bromo-5-chloro-1,1,3,3,5,5-hexafluoropentane
Formel: C₅H₄BrClF₆
Molecular Weight: 293,433 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28901029
CAS-Nummer: 1980045-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR