1-(Perfluoropentyl)pentane 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,1,1,2,2,3,3,4,4,5,5-Undecafluorodecane

PC56224-1G
APOSPC56224-1GEA 139 EUR
APOSPC56224-1G
1-(Perfluoropentyl)pentane 97%
1-(Perfluoropentyl)pentane
Formel: C₁₀H₁₁F₁₁
Molecular Weight: 340,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28135712
CAS-Nummer: 1287702-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR