2-(2,2,3,4,4,4-Hexafluorobutoxy)ethanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2,2,3,4,4,4-Hexafluorobutoxy)ethanol
PC56226-5G
APOSPC56226-5GEA
79.9
EUR
APOSPC56226-5G
2-(2,2,3,4,4,4-Hexafluorobutoxy)ethanol
2-(2,2,3,4,4,4-Hexafluorobutoxy)ethanol
Formel:
C₆H₈F₆O₂ Molecular Weight: 226.117 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972891 CAS-Nummer: 63693-10-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...