2-[(4-Bromophenoxy)methyl]-1,1,3,3,3-pentafluoro-1-propene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-4-[3,3-difluoro-2-(trifluoromethyl)prop-2-enoxy]benzene

PC56228-1G
APOSPC56228-1GEA 174 EUR
APOSPC56228-1G
2-[(4-Bromophenoxy)methyl]-1,1,3,3,3-pentafluoro-1-propene
2-[(4-Bromophenoxy)methyl]-1,1,3,3,3-pentafluoro-1-propene
Formel: C₁₀H₆BrF₅O
Molecular Weight: 317,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334477
CAS-Nummer: 1980085-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR