1-Bromo-4-[1,3,3,3-tetrafluoroprop-1-enoxy]benzene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-4-[1,3,3,3-tetrafluoroprop-1-enoxy]benzene

PC56229-1G
APOSPC56229-1GEA 87 EUR
APOSPC56229-1G
1-Bromo-4-[1,3,3,3-tetrafluoroprop-1-enoxy]benzene
1-Bromo-4-[1,3,3,3-tetrafluoroprop-1-enoxy]benzene
Formel: C₉H₅BrF₄O
Molecular Weight: 285,033 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054318
CAS-Nummer: 1824865-81-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR