1H-Perfluoro-2,5-dioxahexane

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(difluoromethoxy)-1,1,2,2-tetrafluoro-2-(trifluoromethoxy)ethane

PC56230-1G PC56230-5G
APOSPC56230-1GEA 212 EUR
APOSPC56230-1G APOSPC56230-5G
1H-Perfluoro-2,5-dioxahexane
1H-Perfluoro-2,5-dioxahexane
Formel: C₄HF₉O₂
Molecular Weight: 252.035 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31580163
CAS-Nummer: 40891-98-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR