1-Bromo-4-{[(Z)-2,3,3,3-tetrafluoroprop-1-en-1-yl]oxy}benzene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-4-[(Z)-2,3,3,3-tetrafluoroprop-1-enoxy]benzene

PC56231-1G
APOSPC56231-1GEA 72.5 EUR
APOSPC56231-1G
1-Bromo-4-{[(Z)-2,3,3,3-tetrafluoroprop-1-en-1-yl]oxy}benzene
1-Bromo-4-{[(Z)-2,3,3,3-tetrafluoroprop-1-en-1-yl]oxy}benzene
Formel: C₉H₅BrF₄O
Molecular Weight: 285,033 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28137307
CAS-Nummer: 1824879-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR