4,4,5,5-Tetrachloro-2,2-difluoro-1,3-dioxolane

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4,4,5,5-Tetrachloro-2,2-difluoro-1,3-dioxolane

PC56232-1G
APOSPC56232-1GEA 323 EUR
APOSPC56232-1G
4,4,5,5-Tetrachloro-2,2-difluoro-1,3-dioxolane
4,4,5,5-Tetrachloro-2,2-difluoro-1,3-dioxolane
Formel: C₃Cl₄F₂O₂
Molecular Weight: 247,84 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054322
CAS-Nummer: 87075-01-2
UN: 1992
ADR: 3+6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR