2-Bromo-4-fluoro-1-(trifluoromethoxy)benzene 98%

Lieferant: Apollo Scientific

PC56241-1G
APOSPC56241-1GEA 134 EUR
APOSPC56241-1G
2-Bromo-4-fluoro-1-(trifluoromethoxy)benzene 98%
2-Bromo-4-fluoro-1-(trifluoromethoxy)benzene
Formel: C₇H₃BrF₄O
Molecular Weight: 258.996 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847187
CAS-Nummer: 866633-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR