Methyl 2-(3-fluoro-4-nitrophenyl)acetate 95%

Lieferant: Apollo Scientific

PC56250-1G
APOSPC56250-1GEA 368 EUR
APOSPC56250-1G
Methyl 2-(3-fluoro-4-nitrophenyl)acetate 95%
Methyl 2-(3-fluoro-4-nitrophenyl)acetate
Formel: C₉H₈FNO₄
Molecular Weight: 213,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16620330
CAS-Nummer: 169339-41-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR