5-(Dimethoxymethyl)-2-fluoroaniline 95%

Lieferant: Apollo Scientific

PC56292-5G
APOSPC56292-5GEA 1180 EUR
APOSPC56292-5G
5-(Dimethoxymethyl)-2-fluoroaniline 95%
5-(Dimethoxymethyl)-2-fluoroaniline
Formel: C₉H₁₂FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD20716984
CAS-Nummer: 1378862-38-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR