2-(Dimethoxymethyl)-4-fluoroaniline 95%

Lieferant: Apollo Scientific

PC56294-5G
APOSPC56294-5GEA 1180 EUR
APOSPC56294-5G
2-(Dimethoxymethyl)-4-fluoroaniline 95%
2-(Dimethoxymethyl)-4-fluoroaniline
Formel: C₉H₁₂FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31614285
CAS-Nummer: 2244087-31-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR