4-(Dimethoxymethyl)-3-(trifluoromethyl)aniline 95%

Lieferant: Apollo Scientific

PC56296-5G
APOSPC56296-5GEA 1320 EUR
APOSPC56296-5G
4-(Dimethoxymethyl)-3-(trifluoromethyl)aniline 95%
4-(Dimethoxymethyl)-3-(trifluoromethyl)aniline
Formel: C₁₀H₁₂F₃NO₂
Molecular Weight: 235.203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31614286
CAS-Nummer: 2244083-50-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR