3-(Dimethoxymethyl)-2-fluoroaniline 95%

Lieferant: Apollo Scientific

PC56299-5G
APOSPC56299-5GEA 1470 EUR
APOSPC56299-5G
3-(Dimethoxymethyl)-2-fluoroaniline 95%
3-(Dimethoxymethyl)-2-fluoroaniline
Formel: C₉H₁₂FNO₂
Molecular Weight: 185,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31614287
CAS-Nummer: 2244087-96-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR