4-(Dimethoxymethyl)-3-fluoroaniline 95%

Lieferant: Apollo Scientific

PC56300-5G
APOSPC56300-5GEA 1180 EUR
APOSPC56300-5G
4-(Dimethoxymethyl)-3-fluoroaniline 95%
4-(Dimethoxymethyl)-3-fluoroaniline
Formel: C₉H₁₂FNO₂
Molecular Weight: 185,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31614288
CAS-Nummer: 1428246-55-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR