4-(Dimethoxymethyl)-2-fluoroaniline 95%

Lieferant: Apollo Scientific

PC56305-5G
APOSPC56305-5GEA 1180 EUR
APOSPC56305-5G
4-(Dimethoxymethyl)-2-fluoroaniline 95%
4-(Dimethoxymethyl)-2-fluoroaniline
Formel: C₉H₁₂FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD20722958
CAS-Nummer: 232950-77-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR