6-Fluoro-2-methyl-3-nitrobenzyl alcohol 97%

Lieferant: Apollo Scientific

Synonyme: (6-Fluoro-2-methyl-3-nitrophenyl)methanol

PC56307-250MG
APOSPC56307-250MGEA 36.4 EUR
APOSPC56307-250MG
6-Fluoro-2-methyl-3-nitrobenzyl alcohol 97%
6-Fluoro-2-methyl-3-nitrobenzyl alcohol
Formel: C₈H₈FNO₃
Molecular Weight: 185,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28347725
CAS-Nummer: 1613329-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR