Sodium tetrakis(4-fluorophenyl)borate dihydrate 99+%
Lieferant: Apollo Scientific
|
Achtung
|
PC56331-5G
PC56331-1G
APOSPC56331-5GEA
957
EUR
APOSPC56331-5G
APOSPC56331-1G
Sodium tetrakis(4-fluorophenyl)borate dihydrate 99+%
Sodium tetrakis(4-fluorophenyl)borate dihydrate
Formel:
C₂₄H₂₀BF₄NaO₂ Molecular Weight: 450.215 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00149598 CAS-Nummer: 207683-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...