Sodium tetrakis(4-fluorophenyl)borate dihydrate 99+%

Lieferant: Apollo Scientific
Achtung

PC56331-5G PC56331-1G
APOSPC56331-5GEA 957 EUR
APOSPC56331-5G APOSPC56331-1G
Sodium tetrakis(4-fluorophenyl)borate dihydrate 99+%
Sodium tetrakis(4-fluorophenyl)borate dihydrate
Formel: C₂₄H₂₀BF₄NaO₂
Molecular Weight: 450.215 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00149598
CAS-Nummer: 207683-22-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR