6-Bromo-3-fluoro-2-nitrophenol 95%

Lieferant: Apollo Scientific
Achtung

PC56337-1G
APOSPC56337-1GEA 163 EUR
APOSPC56337-1G
6-Bromo-3-fluoro-2-nitrophenol 95%
6-Bromo-3-fluoro-2-nitrophenol
Formel: C₆H₃BrFNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD23701729
CAS-Nummer: 872363-59-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR