3-(Trifluormethyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Trifluormethyl)benzolboronsäure, Dihydroxy(3-(trifluormethyl)phenyl)boran, (3-(Trifluormethyl)phenyl)metaborsäure

PC5641-100G PC5641-25G PC5641-5G
APOSPC5641-100GEA 132 EUR
APOSPC5641-100G APOSPC5641-25G APOSPC5641-5G
3-(Trifluormethyl)phenylboronsäure 98%
3-(Trifluormethyl)phenylboronsäure
Formel: C₇H₆BF₃O₂
Molecular Weight: 189.93 g/mol
Schmelzpunkt: 162…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151854
CAS-Nummer: 1423-26-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR