3-(Trifluormethyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Trifluormethyl)benzolboronsäure, Dihydroxy(3-(trifluormethyl)phenyl)boran, (3-(Trifluormethyl)phenyl)metaborsäure
PC5641-100G
PC5641-25G
PC5641-5G
APOSPC5641-100GEA
132
EUR
APOSPC5641-100G
APOSPC5641-25G
APOSPC5641-5G
3-(Trifluormethyl)phenylboronsäure 98%
3-(Trifluormethyl)phenylboronsäure
Formel:
C₇H₆BF₃O₂ Schmelzpunkt: 162…164 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151854 CAS-Nummer: 1423-26-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...