4-(Trifluoromethyl)benzamidine hydrochloride dihydrate 97%

Lieferant: Apollo Scientific
Achtung

PC5648-1G
APOSPC5648-1GEA 21.9 EUR
APOSPC5648-1G
4-(Trifluoromethyl)benzamidine hydrochloride dihydrate 97%
4-(Trifluoromethyl)benzamidine hydrochloride dihydrate
Formel: C₈H₁₂ClF₃N₂O₂
Schmelzpunkt: 155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00122370
CAS-Nummer: 175278-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR