2-Brom-5-methylbenzotrifluorid 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC5649-1G
APOSPC5649-1GEA
21.9
EUR
APOSPC5649-1G
2-Brom-5-methylbenzotrifluorid 98%
2-Brom-5-methylbenzotrifluorid
Formel:
C₈H₆BrF₃ Molecular Weight: 239,04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631586 CAS-Nummer: 261952-20-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...