Bis(4-fluoro-3-nitrophenyl)sulphone

Lieferant: Apollo Scientific

Synonyme: DFDNPS, 4,4'-Difluoro-3,3'-dinitrodiphenyl Sulphone

PC5662-5G
APOSPC5662-5GEA 132 EUR
APOSPC5662-5G
Bis(4-fluoro-3-nitrophenyl)sulphone
Bis(4-fluoro-3-nitrophenyl)sulphone
Formel: C₁₂H₆F₂N₂O₆S
Schmelzpunkt: 194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007057
CAS-Nummer: 312-30-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR