3-(Trifluormethoxy)phenylboronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

PC5663-1G PC5663-5G
APOSPC5663-1GEA 0 EUR
APOSPC5663-1G APOSPC5663-5G
3-(Trifluormethoxy)phenylboronsäurepinakolester
3-(Trifluormethoxy)phenylboronsäurepinakolester
Formel: C₁₃H₁₆BF₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05863919
CAS-Nummer: 262376-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR