Ethyl 4,4-difluoro-2-(ethoxymethylene)acetoacetate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-(ethoxymethylene)-4,4-difluoro-3-oxobutanoate, Ethyl 4,4-difluoro-2-(ethoxymethylene)-3-oxobutanoate, Ethyl 4,4-difluoro-2-(ethoxymethylidene)-3-oxobutanoate
PC5667-1G
PC5667-5G
APOSPC5667-1GEA
190
EUR
APOSPC5667-1G
APOSPC5667-5G
Ethyl 4,4-difluoro-2-(ethoxymethylene)acetoacetate
Ethyl 4,4-difluoro-2-(ethoxymethylene)acetoacetate
Formel:
C₉H₁₂F₂O₄ Siedepunkt: 85…87 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD10567161 CAS-Nummer: 176969-33-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...