Ethyl 4,4-difluoro-2-(ethoxymethylene)acetoacetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-(ethoxymethylene)-4,4-difluoro-3-oxobutanoate, Ethyl 4,4-difluoro-2-(ethoxymethylene)-3-oxobutanoate, Ethyl 4,4-difluoro-2-(ethoxymethylidene)-3-oxobutanoate

PC5667-1G PC5667-5G
APOSPC5667-1GEA 190 EUR
APOSPC5667-1G APOSPC5667-5G
Ethyl 4,4-difluoro-2-(ethoxymethylene)acetoacetate
Ethyl 4,4-difluoro-2-(ethoxymethylene)acetoacetate
Formel: C₉H₁₂F₂O₄
Siedepunkt: 85…87 °C
Lagertemperatur: Kühlschrank
MDL: MFCD10567161
CAS-Nummer: 176969-33-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR