1-Chloro-2,3,3,3-tetrafluoropropene

Lieferant: Apollo Scientific
Achtung

PC56700-5G
APOSPC56700-5GEA 691 EUR
APOSPC56700-5G
1-Chloro-2,3,3,3-tetrafluoropropene
1-Chloro-2,3,3,3-tetrafluoropropene
Formel: C₃HClF₄
Molecular Weight: 148,487 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30062788
CAS-Nummer: 3110-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR