(Z)-1-Chloro-2,3,3,3-tetrafluoropropene 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC56702-5G
APOSPC56702-5GEA
522
EUR
APOSPC56702-5G
(Z)-1-Chloro-2,3,3,3-tetrafluoropropene 97%
(Z)-1-Chloro-2,3,3,3-tetrafluoropropene
Formel:
C₃HClF₄ Molecular Weight: 148,487 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30062788 CAS-Nummer: 111512-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...