4-Amino-N-(4-trifluoromethylphenyl)benzamide 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC56728-250MG
APOSPC56728-250MGEA
580
EUR
APOSPC56728-250MG
4-Amino-N-(4-trifluoromethylphenyl)benzamide 95%
4-Amino-N-(4-trifluoromethylphenyl)benzamide
Formel:
C₁₄H₁₁F₃N₂O Molecular Weight: 280,245 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09802491 CAS-Nummer: 1011244-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...