2,4-Difluoro-3-(trifluoromethoxy)phenylacetylene 95%

Lieferant: Apollo Scientific
Achtung

PC56733-1G PC56733-250MG
APOSPC56733-1GEA 0 EUR
APOSPC56733-1G APOSPC56733-250MG
2,4-Difluoro-3-(trifluoromethoxy)phenylacetylene 95%
2,4-Difluoro-3-(trifluoromethoxy)phenylacetylene
Formel: C₉H₃F₅O
Lagertemperatur: Kühlschrank
MDL: MFCD31618862
CAS-Nummer: 2237235-34-4
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR