3-Chloro-5-fluorophenylacetylene 95+%

Lieferant: Apollo Scientific

Synonyme: 1-Chloro-3-ethynyl-5-fluorobenzene

PC56740-1G
APOSPC56740-1GEA 1180 EUR
APOSPC56740-1G
3-Chloro-5-fluorophenylacetylene 95+%
3-Chloro-5-fluorophenylacetylene
Formel: C₈H₄ClF
Molecular Weight: 154,569 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11618081
CAS-Nummer: 1233506-04-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR