2-Chloro-2,2-difluoro-1-(4-fluorophenyl)ethanone 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC56748-1G
PC56748-250MG
APOSPC56748-1GEA
654
EUR
APOSPC56748-1G
APOSPC56748-250MG
2-Chloro-2,2-difluoro-1-(4-fluorophenyl)ethanone 95%
2-Chloro-2,2-difluoro-1-(4-fluorophenyl)ethanone
Formel:
C₈H₄ClF₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD20921049 CAS-Nummer: 1321028-35-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...