2-Chloro-4-(trifluoromethoxy)benzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

PC56780-1G PC56780-5G
APOSPC56780-1GEA 873 EUR
APOSPC56780-1G APOSPC56780-5G
2-Chloro-4-(trifluoromethoxy)benzyl bromide 95%
2-Chloro-4-(trifluoromethoxy)benzyl bromide
Formel: C₈H₅BrClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD18393829
CAS-Nummer: 1261642-94-7
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR