6-Bromo-3-(difluoromethyl)-1H-indole, N-BOC protected
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
tert-butyl 6-bromo-3-(difluoromethyl)indole-1-carboxylate
PC56795-100MG
PC56795-1G
PC56795-500MG
APOSPC56795-100MGEA
522
EUR
APOSPC56795-100MG
APOSPC56795-1G
APOSPC56795-500MG
6-Bromo-3-(difluoromethyl)-1H-indole, N-BOC protected
6-Bromo-3-(difluoromethyl)-1H-indole, N-BOC protected
Formel:
C₁₄H₁₄BrF₂NO₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
2237235-37-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...