2-(2,6-Difluorophenyl)propionic acid 95%

Lieferant: Apollo Scientific
Achtung

PC56800-1G PC56800-250MG
APOSPC56800-1GEA 1010 EUR
APOSPC56800-1G APOSPC56800-250MG
2-(2,6-Difluorophenyl)propionic acid 95%
2-(2,6-Difluorophenyl)propionic acid
Formel: C₉H₈F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11036843
CAS-Nummer: 359828-68-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR