3-[2-Nitro-4-(trifluoromethyl)phenoxy]benzeneboronic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC5681-1G
APOSPC5681-1GEA
184
EUR
APOSPC5681-1G
3-[2-Nitro-4-(trifluoromethyl)phenoxy]benzeneboronic acid
3-[2-Nitro-4-(trifluoromethyl)phenoxy]benzeneboronic acid
Formel:
C₁₃H₉BF₃NO₅ Molecular Weight: 327,02 g/mol Schmelzpunkt: 111…113 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09972104 CAS-Nummer: 1072945-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...