2-Bromo-4-(trifluoromethyl)phenylacetylene 95%

Lieferant: Apollo Scientific
Achtung

PC56840-1G PC56840-250MG
APOSPC56840-1GEA 574 EUR
APOSPC56840-1G APOSPC56840-250MG
2-Bromo-4-(trifluoromethyl)phenylacetylene 95%
2-Bromo-4-(trifluoromethyl)phenylacetylene
Formel: C₉H₄BrF₃
Lagertemperatur: Kühlschrank
MDL: MFCD31689284
CAS-Nummer: 871126-88-4
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR