3-Bromo-5-(trifluoromethyl)phenylacetylene 95%

Lieferant: Apollo Scientific
Achtung

PC56842-1G PC56842-250MG
APOSPC56842-1GEA 0 EUR
APOSPC56842-1G APOSPC56842-250MG
3-Bromo-5-(trifluoromethyl)phenylacetylene 95%
3-Bromo-5-(trifluoromethyl)phenylacetylene
Formel: C₉H₄BrF₃
Lagertemperatur: Kühlschrank
MDL: MFCD21243959
CAS-Nummer: 1494947-64-6
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR