3,6-Difluor-2-methoxyphenylboronsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3,6-Difluoro-2-methoxyphenyl)boronic acid
PC56845-1G
PC56845-250MG
PC56845-5G
APOSPC56845-1GEA
455
EUR
APOSPC56845-1G
APOSPC56845-250MG
APOSPC56845-5G
3,6-Difluor-2-methoxyphenylboronsäure 95%
3,6-Difluor-2-methoxyphenylboronsäure
Formel:
C₇H₇BF₂O₃ Molecular Weight: 187.936 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD11856009 CAS-Nummer: 919355-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...